Flasks, pear shaped, acc. to Claisen, with two necks
Leverandør: witeg Labortechnik
0.783.025
WITG0.783.025EA 2250 NOK
WITG0.783.025
Flasks, pear shaped, acc. to Claisen, with two necks
Kolber Pæreformet flaske
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Center necks with socked NS 14/23, side neck with cone NS 14/23.