Flasks, pear shaped, Claisen-Vigreux, with two necks
Leverandør: witeg Labortechnik
0.786.025
WITG0.786.025EA 2900 NOK
WITG0.786.025
Flasks, pear shaped, Claisen-Vigreux, with two necks
Kolber Pæreformet flaske
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Incorporates a short, Vigreux type, fractionating column. Column indents aid intimate contact between vapour and liquid.