Funnels, with standard ground joint cone
Leverandør: witeg Labortechnik
1.650.014 1.650.029 1.650.129 1.650.130 1.650.150 1.650.019
WITG1.650.014EA 265 NOK
WITG1.650.014 WITG1.650.029 WITG1.650.129 WITG1.650.130 WITG1.650.150 WITG1.650.019
Funnels, with standard ground joint cone
Trakter Filtreringstrakter Trakter med konisk NS-slip
Funnels, made of boroslicate glass, with standard ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.