Dropping funnels, cylindrical, without pressure balance
Leverandør: witeg Labortechnik
1.856.002
WITG1.856.002EA 8400 NOK
WITG1.856.002
Dropping funnels, cylindrical, without pressure balance
Trakter Dråpetrakter
Dropping funnels, made of borosilicate glass 3.3, cylindrical, stopcock with glass key and screw-thread retaling nut.
  • Graduated
  • With NS cone and socket
  • Very good chemical and temperature resistance
Sertifiseringer: DIN 12567