Dosing funnels, with spiral conveyor
Leverandør: witeg Labortechnik
1.897.825
WITG1.897.825EA 17825 NOK
WITG1.897.825
Dosing funnels, with spiral conveyor
Trakter Dråpetrakter
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For powdery substances. Horizontal version with spiral conveyor.