Adapters for distilling receivers
Leverandør: witeg Labortechnik
1.920.001
WITG1.920.001EA 1375 NOK
WITG1.920.001
Adapters for distilling receivers
Destillasjonsutstyr Destillasjonsadaptere
Borosilicate glass 3,3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes