Gas generators, Duran®
Leverandør: witeg Labortechnik
2.989.500
WITG2.989.500EA 10125 NOK
WITG2.989.500
Gas generators, Duran®
Gassylindre
Gas generators according to Kipp, Duran.
  • Tubulature at bottom
  • Jointless, total grease free apparatus
With two screw threads GL 25, one screw thread GL 45 and GL 18, with needle valve stopcock with overtwist device.
Leveringsinformasjon: Gas generators complete with safety funnel, caps and seals, hole disc made of sterilisable silicone, 500 ml capacity of middle bulb.