Culture tubes
Leverandør: witeg Labortechnik
4.903.101
WITG4.903.101EA 470 NOK
WITG4.903.101
Culture tubes
Rør Kulturrør
Borosilicate glass 3,3 with rim.
  • Excellent chemical resistance
  • Excellent temperature resistance