Begerglass, høy modell
Leverandør: witeg Labortechnik
5.503.500
WITG5.503.500EA 1275 NOK
WITG5.503.500
Begerglass, høy modell
Beger
Borosilicate glass 3.3, with handle.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for general laboratory use.