Flasks acc. to Kjeldahl, with beaded rim
Leverandør: witeg Labortechnik
5.518.001
WITG5.518.001EA 1125 NOK
WITG5.518.001
Flasks acc. to Kjeldahl, with beaded rim
Kolber Kjeldahlkolber
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With printed indication of contents and white labelling area.