Distillation flasks, engler type
Leverandør: witeg Labortechnik
5.522.125
WITG5.522.125EA 760 NOK
WITG5.522.125
Distillation flasks, engler type
Kolber Destillasjonskolber
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.