Woulff bottles
Leverandør: witeg Labortechnik
5.663.000
WITG5.663.000EA 6750 NOK
WITG5.663.000
Woulff bottles
Flasker Woulff-flasker
Glass, clear, plastic coated.
  • Clear plastic coated to protect against mechanical breakage
  • Reduces the hazard of flying glass and liquid splashes in the event of breakage
  • Implosion protection
Complete set includes 500 ml bottle with 3 necks (center neck GL 25, side necks GL 18), suction tube with safety valve, analog pressure gauge. Air-suction tube icnludes PTFE needle valve stopcock.