Density bottles, adjusted

Leverandør: witeg Labortechnik
8.126.139
WITG8.126.139EA 1575 NOK
WITG8.126.139
Density bottles, adjusted
Pyknometer
Pycnometer, according to Hubbard

  • Adjusted to second decimal place

For use with bitumen and heavy oils.

Sertifiseringer: According to DIN 12806 and ASTM D-70
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR