Reciprocating shaker, SHR digital
Leverandør: witeg Labortechnik
DH.WSR04022
WITGDH.WSR04022EA 27850 NOK
WITGDH.WSR04022
Reciprocating shaker, SHR digital
Risteapparater Risteapparater, sideveis
These digital reciprocating shakers with programmable digital feedback control system are ideal for culturing cells in hybridisation, staining and destaining gels. Also used in combinatorial chemistry.
  • Brushless DC motor (low noise and long life cycle)
  • High precision digital rotation speed control
  • Over-Load protection, storage function and alarm
  • Programmable digital feedback controller with Jog-Shuttle switch (turn + push)
The optimal platform rotation helps for intensive mixing with convenient setting of timer and speed using jog shuttle switch. ForMore Product Information
Bestill nå
SPESIFIKASJONER
Modell SHR-2D
Ristebevegelse Reciprocating
Environmentally Preferable
Amplitude 30 mm
Hastighetsområde 10 - 300 min⁻¹
Display Digital LCD
Plattform B × D 350×350 mm
B×D×H 400×480×160 mm
Vekt 20 kg
Maks. last 10 kg