Wright's stain modified, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

WS32-1L
WS32-1LEA 1800 NOK
WS32-1L
Wright's stain modified, Sigma-Aldrich®
Wright's stain

Popular hematology stain used for differentially staining the cellular elements of blood. For dip, rack, and batch staining techniques.

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082143
CAS-nummer: 68988-92-1
ADR: 3 III
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR