MES sodium salt ≥98%, Reagent Grade

Leverandør: VWR Chemicals

Synonymer: MES-Na

X218-500GEA 8050 NOK
X218-500G
MES sodium salt ≥98%, Reagent Grade
MES sodium salt
Formel: C₆H₁₂NNaO₄S
MW: 217.22 g/mol
MDL nummer: MFCD00065473
CAS-nummer: 71119-23-8
EINECS: 275-203-2

Bestill nå

Spesifikasjon Testresultater

Appearance White crystalline powder
Moisture ≤10%
pH (1% Water) @ 25 °C 9 − 10.5
Purity ≥98%
Solubility (10%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR