Polysorbate 20, J.T.Baker®

Leverandør: Avantor

X251-07EA 4550 NOK
X251-07 X251-09
Polysorbate 20, J.T.Baker®
Polysorbate 20
Formel: C₅₈H₁₁₄O₂₆
Tetthet: 1,1 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00165986
CAS-nummer: 9005-64-5
EINECS: 500-018-3

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use Passes Test
Identification (by IR) Passes Test
Not intended for Injectable Drug Use.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR