Polysorbat 80, J.T.Baker®

Leverandør: Avantor

X257-09EA 0 NOK
X257-09 X257-01
Polysorbat 80, J.T.Baker®
Polysorbat 80

Polyoxyethylene (20) Sorbitan Monooleate

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082107
CAS-nummer: 9005-65-6
EINECS: 500-018-3

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use Passes Test
Identification (by IR) Passes Test
Not intended for Injectable Drug Use.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR