Multi-parameter meter, handheld, Pro 1030
Leverandør: YSI ENVIRONMENTAL
6051030
YSIA6051030EA 19275 NOK
YSIA6051030
Multi-parameter meter, handheld, Pro 1030
Elektrokjemiske målere pH-meter
This rugged and reliable instrument is suitable for the measurement of either pH or ORP along with conductivity and temperature. Milspec (military spec) connectors, IP 67 waterproof and rubber over moulded case, and 1-meter drop tests ensure the instrument provides years of sampling even in the harshest field conditions.
  • User-replaceable cables and sensors (pH and ORP); stable 4-electrode conductivity sensor built into the cable
  • Smart calibration in the instrument remembers the previous calibration values and simply walks you through the routine
  • Stores 50