Ice Makers CoolNat Propane (R290)
Leverandør: ZIEGRA EISMASCHINEN
1001000301VPCH 1001000301VPUK 1001000703VPCH 1001000703VPUK 1001001103VPCH 1001001103VPUK 1001000700VPCH 1001000700VPUK 1001000701VPCH 1001000701VPUK 1001001500VPCH 1001001500VPUK 1001001501VPCH 1001001501VPUK 1001001502VPCH 1001001502VPUK 1001002500VPCH 1001002500VPUK 1001002501VPCH 1001002501VPUK 1001002502VPCH 1001002502VPUK 8400015101114
474-0140EA 56150 NOK
474-0140 474-0142 474-0150 474-0152 474-0154 474-0156 474-0143 474-0145 474-0146 474-0148 474-0157 474-0159 474-0160 474-0162 474-0163 474-0165 474-0166 474-0168 474-0169 474-0171 474-0172 474-0174 ZIEG8400015101114
Ice Makers CoolNat Propane (R290)
Ismaskiner
Ice machines for small and moderate requirements with storage and other accessories, this extensive range of propane (R290) ice machines offers solutions for all applications. Ice machines in the CoolNat propane series come equipped with high quality safety valves and the most up-to-date pressure monitors to ensure the safest possible use of propane. Minimal amounts of refrigerant are used, well within the levels set by the most stringent of technical safety requirements. The CoolNat range offers the same features than the standard serie.
  • Choice of micro or standard ice sizes ensure