Multitex® Wipes, Loose Wiping Cloth
Leverandør: Zellstoff-Vertriebs-GmbH & Co. KG
10840-00
ZVGA10840-00EA 1075 NOK
ZVGA10840-00
Multitex® Wipes, Loose Wiping Cloth
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
Non-woven wipes for disinfection and cleaning.
  • 100% PET
  • Premium smooth
Tear-resistant, absorbent and low-lint.