zetMedica®/zetMask® Disposable Face Masks
Leverandør: Zellstoff-Vertriebs-GmbH & Co. KG
30960-01
ZVGA30960-01EA 3850 NOK
ZVGA30960-01
zetMedica®/zetMask® Disposable Face Masks
Masker Munnbind, renrom
zetMedica®/zetMask® disposable face mask combines high bacterial filter efficiency with optimal wearing comfort and
protects the sensitive skin of the mouth and nose. A wide variety of shapes and materials have proven themselves in the care sector as well as in the high-tech and food industries.
  • Nonwoven face mask, with elastic band
The face mask is fiberglass-free, latex-free, suitable for people who wear glasses and is available in a practical dispenser box.