HPLC columns, Acclaim™ 300 C18
Leverandør: Thermo Fisher Scientific
069690 069697 063684 060263 060264 060265 060266
555-7118EA 3725 NOK
555-7118 555-7131 555-7299 555-7312 555-7313 555-7314 555-7315
HPLC columns, Acclaim™ 300 C18
Kromatografikolonner
These high resolution reversed phase columns are used to separate proteins and peptides.
  • High efficiency 3 µm spherical silica substrate
  • Application tested suitable for peptide mapping
  • Reproducible for dependable results
  • LC/MS compatible
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning
Kappet ved endene
pH område
Overflate areal
Temperaturgrense
USP kode