HPLC columns, MAbPac™ HIC-Butyl
Leverandør: Thermo Fisher Scientific
088558 088559
555-7360EA 16325 NOK
555-7360 555-7361
HPLC columns, MAbPac™ HIC-Butyl
Kromatografikolonner
The MAbPac™ HIC-Butyl HPLC columns are used to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). The column's hydrophilic resin provides excellent biocompatibility and low carryover, while the optimal density of the butyl groups provides high resolution separation.
  • High resolution and high efficiency
  • Stable at wide range of pH (2 to 12)
  • Low carryover
  • Rugged column packing
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. trykk 4000 psi
pH område 2 - 12
Temperaturgrense 60 °C