LC columns, MAbPac™ RP
Leverandør: Thermo Fisher Scientific
088644 088645 088646 088647 088648 088649 303182 303183 303184 303270
554-4732EA 10775 NOK
554-4732 554-4733 554-4734 554-4735 554-4736 554-4737 554-6580 554-6581 554-6582 554-6583
LC columns, MAbPac™ RP
Kromatografikolonner
These columns are designed for high performance reversed-phase HPLC and LC-MS characterisation of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins and bispecific proteins.
  • Efficient separation of protein molecules with very low carry over
  • Excellent MS compatibility
  • High throughput
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. trykk 4000 psi
pH område 0 - 14
Temperaturgrense <110 °C