Masterflex® L/S® Precision Pump Tubing, Peroxide-Cured Silicone, L/S 25; 25 ft
Leverandør: Avantor Fluid Handling
VWR Item#MFLX96400-25
Your Organization's Price:
Excellent biocompatibility for a wide range of applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Greater physical compression capability
  • Tubing Size: L/S 25
<B>Applications</B>: Pharmaceutical, biotech, peristaltic pumping, and food applications<br /><B>Characteristics</B>: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.