Stainless Steel (SS) Hose Clamps, 7/32" x 5/8"; 10/Pk
Leverandør: Avantor Fluid Handling
VWR Item#MFLX06403-11
Your Organization's Price:
Ideal for securely clamping tubes in place.
  • Resists corrosion
  • Clamp size: 1-5/16" ID x 2-1/4" ID
The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.