Masterflex® Hose Clamp, Acetal Copolymer, White, 0.779" ID x 0.889" ID; 100/PK
Leverandør: Avantor Fluid Handling
VWR Item#MFLX06832-18
Your Organization's Price:
Reusable white acetal copolymer clamps will not rust or corrode.
  • Resists corrosion
  • Clamp size: 0.779" ID x 0.889" ID
Recommended for continuous use from ambient temperature to 200ºF (93ºC). Clamps are nonconducting and stable in a wide variety of chemicals. <br /><br />Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers.
The acetal hose clamp kit includes 170 assorted clamps from 0.228" to 1.405" ID.