Masterflex® L/S® Standard Pump Head Replacement Thrust Washers; 10/PK
Leverandør: Avantor Fluid Handling
VWR Item#MFLX07021-04
Your Organization's Price:
Get the most out of your Masterflex® L/S® Standard Pump Head with these replacement parts.
  • Suitable for use with Masterflex® L/S® Standard pump heads
Replacement parts to keep your pump head operating even longer. Thrust washers fit on either side of the rotor assembly of L/S standard pump heads.