Masterflex® Transfer Tubing, Tygon® A-60-F I.B., 1/4" ID x 1/2" OD; 50 Ft
Leverandør: Avantor Fluid Handling
VWR Item#MFLX95715-00
Your Organization's Price:
Flexibility at extreme temperatures.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 1/4" ID x 1/2" OD
Ideal for chemical transfer, pesticide/fertilizer delivery, water feed lines, food/dairy processing, and water purification lines. The tube features excellent resistance to acidic and alkali substances, ozone, UV light, and abrasive media. It is nonconductive, heat-sealable, non-aging, and nonoxidizing. In addition, the tube maintains flexibility even at extreme temperatures. Temperature range is -60 to 275 °F (-51 to 135 °C). Can be sterilized with autoclave.