Double-walled PTFE heat-shrink tubing, 0.180"ID, 12 ft/pack
Leverandør: Avantor Fluid Handling
VWR Item#MFLX95810-13
Your Organization's Price:
Ideal for chemically inert applications.
  • Global market leader in range of formulations for most extensive chemical compatibility
Double-walled PTFE tubing combines an exterior of heat-shrink PTFE with an inner layer of FEP to provide a tight, moisture-proof bond. Comes in 4-ft lengths; 3 lengths per pack, for a total of 12 ft/pk. Opaque.