Adapter for 1/4" ID Flexible Tubing, Stainless Steel
Leverandør: Avantor Fluid Handling
VWR Item#MFLX07104-81
Your Organization's Price:
Ideal adapters for plastic case pump heads.
  • Optimize performance of your FMI RH Series Piston Pumps
Adapters for your FMI RH series piston pump heads.