Micropump NEMA 56C Cavity Style Pump Head; 100psi/SS/PEEK/TEV; 3.48 mL/rev
Leverandør: Avantor Fluid Handling
VWR Item#MFLX73007-02
Your Organization's Price:
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Compact size and easy to service
  • Cavity-style heads are designed to handle higher inlet pressures.
Magnetically driven, precision-geared cavity pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and a single seal keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of fluids.