Suction Line Strainer, PVC, 1-1/2" NPT(F), 80 Mesh (stainless steel)
Leverandør: Avantor Fluid Handling
VWR Item#MFLX74604-54
Your Organization's Price:
Protect pump from sediments filtering into suction line.
  • PVC (polyvinyl chloride) or Nylon base material of construction
  • 20, 40, 80 mesh sizes available
  • Easily removable for cleaning or replacing
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.