Masterflex® Transfer Tubing, Polyimide, 0.0049" ID x 0.0064" OD, 12"L; 5/PK
Leverandør: Avantor Fluid Handling
VWR Item#MFLX95820-00
Your Organization's Price:
Provides excellent temperature stability.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 0.0049" ID x 0.0064" OD
The tubing is ideal for use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers. It offers excellent chemical, heat, radiation, and abrasion resistance and high dielectric strength. Resin meets USP Class VI requirements and NEMA standards. The tube has a temperature range of –418 to 896 °F (–250 to 480 °C). Can be sterilized with autoclave, ethylene oxide or gamma irradiation.