Masterflex® Replacement Tubing Set, Viton®, 0.8, 1.6, and 3.0 mm ID
Leverandør: Avantor Fluid Handling
VWR Item#MFLX73160-58
Your Organization's Price:
Choose tubing based on chemical compatibility, flow rate, and your current pump.
  • Global market leader in range of formulations for most extensive chemical compatibility
  • For use with 73160-00 thru 73160-25
Choose from silicone, norprene and viton replacement tubing for your peristaltic pump.