Masterflex® Fitting Clamp, PVDF, Sanitary Tri-Clamp®, 2"; 1/EA
Leverandør: Avantor Fluid Handling
VWR Item#MFLX31811-02
Your Organization's Price:
Maintain sanitary conditions in your processes.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Sanitary Tri-Clamp holds together two sanitary fittings that are 2"
These sanitary clamps are compatible with Tri-Clamp® fittings. The PVDF material has excellent chemical resistance with low leachable characteristics.