Smiths Medical® Fitting, Polycarbonate, Five Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 50/PK
Leverandør: Avantor Fluid Handling
VWR Item#MFLX30600-43BLK
Your Organization's Price:
Regulate flow without any leaking.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Five ports with female luer locks
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
<B>Warning:</B> This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.