Membrane filter funnels, magnetic, Sentino®
Leverandør: Cytiva (Formerly Pall Lab)
4271 4273 4277
516-0437EA 3400 NOK
516-0437 516-0438 516-0439
Membrane filter funnels, magnetic, Sentino®
Trakter Membranfiltertrakter, trakter
Polyphenylsulfone, autoclavable.
  • Accepts 47 mm membrane filters
  • Leakproof magnetic seal allows one-handed operation
  • Graduated at 50 ml increments for accurate sample measurement
Designed for use with the Sentino® microbiology pump.
Effective filtration area: 150 and 300 ml: 9,6 cm²; 500 ml: 13,1 cm²
Overall Ø×H: 150 ml: 76×118 mm; 300 ml: 76×168 mm; 500 ml: 89×166 mm