Funnels, mini
Leverandør: BOCHEM
8858 8857 8856 8855
BOCH8858EA 145 NOK
BOCH8858 BOCH8857 BOCH8856 BOCH8855
Funnels, mini
Trakter Filtreringstrakter Filtertrakter
Mini funnel, made of 18/10 stainless steel.
  • Excellent temperature resistance
  • Good chemical resistance
  • Easy to clean