Microplate racks for revco UxF and HERAfreeze® HFU T freezers
Leverandør: Thermo Scientific
398335 398323 398322 398321 398333 398334 1950639 1950522
HERA398335EA 2875 NOK
HERA398335 HERA398323 HERA398322 HERA398321 HERA398333 HERA398334 HERA1950639 HERA1950522
Microplate racks for revco UxF and HERAfreeze® HFU T freezers
Stativer for rør Mikroplatestativer
Maximise the storage space in your ultra-low temperature freezer with these Thermo Scientific™, racks for Revco™ ExF, DxF and HERAfeeze™ HFU B freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.