Pipette fillers, manual, Profillette™ 406
Leverandør: Socorex Isba
406.002 406.025 406.300 406.010
SOCO406.002EA 295 NOK
SOCO406.002 SOCO406.025 SOCO406.300 SOCO406.010
Pipette fillers, manual, Profillette™ 406
Pipetter Pipettefyllere
Low cost, easy to use pipette fillers suitable for graduated glass and plastic pipettes.
  • For one-hand pipetting
  • Easy filling and dispensing: Rotate wheel to aspirate
  • Fast release by pressing air inlet valve button, or gradual release by gently turning wheel
  • Air inlet valve for fast distribution
  • Colour coding per size
  • Maintenance free