Volumetric flasks, class B, PP
Leverandør: VITLAB
676891 675891
VITL676891EA 395 NOK
VITL676891 VITL675891
Volumetric flasks, class B, PP
Kolber Målekolber
PP, highly transparent, with PP screw cap.
  • With ring mark individually calibrated to ‘In’
  • Thermal stress up to 60 °C does not permanently exceed the tolerance limits
  • To preserve markings, cleaning at no higher than 60 °C is recommended
Class B tolerances according to DIN EN ISO 1042