HPLC columns, Accucore™ Polar Premium
Leverandør: Thermo Fisher Scientific
28026-012105 28026-052130 28026-053030 28026-054630 28026-102130 28026-103030 28026-104630 28026-152130 28026-153030 28026-154630 28026-252130
554-2347EA 7925 NOK
554-2347 554-2296 554-2297 554-2298 554-2299 554-2300 554-2301 554-2302 554-2303 554-2304 554-2305
HPLC columns, Accucore™ Polar Premium
Kromatografikolonner
Accucore™ Polar Premium is a rugged amide embedded C18 phase that offers complementary selectivity to conventional C18. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub-2 µm particles.
  • Complementary selectivity to conventional C18
  • Low back pressure
  • Stable over a wide pH range and compatible with 100% aqueous mobile phase
Bestill nå
SPESIFIKASJONER
Beskrivelse 10039-010 10039-008 10039-006 10039-004 10039-002 10039-000 10038-998 10038-996 10038-696 10038-694
Environmentally Preferable
Karbonbelastning 8% 8% 8% 8% 8% 8% 8% 8% 8% 8%
pH område 1,5 - 10 1,5 - 10 1,5 - 10 1,5 - 10 1,5 - 10 1,5 - 10 1,5 - 10 1,5 - 10 1,5 - 10 1,5 - 10
Overflate areal 90 m²/g 90 m²/g 90 m²/g 90 m²/g 90 m²/g 90 m²/g 90 m²/g 90 m²/g 90 m²/g 90 m²/g