TLC plates, silica gel 60, unmodified layers, DURASIL
Leverandør: MACHEREY-NAGEL
812003 812004 812005 812006 812007 812008 812005.200
552-0260EA 5375 NOK
552-0260 552-0261 552-0262 552-0264 552-0265 552-0266 552-0263
TLC plates, silica gel 60, unmodified layers, DURASIL
TLC-plater
These unmodified standard silica layers for TLC are available as glass plates with or without fluorescent indicator.
  • Hard, water-resistant and wettable layers due to a special binder system
  • More polar than SIL G
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm.