3-Amino-5-phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 3-Amino-5-phenylpyrazole
A2102-1G A2102-5G
TCIAA2102-1GEA 325 NOK
TCIAA2102-1G TCIAA2102-5G
3-Amino-5-phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Amino-5-phenyl-1H-pyrazole
Formel: C₉H₉N₃
MW: 159,19 g/mol
Smeltepunkt: 126°C
MDL nummer: MFCD00191749
CAS-nummer: 1572-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b14fcf4c6830a0dc4fe55acb73c33ff89006bf97&vid=b14fcf4c6830a0dc4fe55acb73c33ff89006bf97&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 125.0 to 129.0 deg-C