Methyl isocyanoacetate 95% tech.

Leverandør: Thermo Scientific
Danger

219000050 219000250
ACRO219000050EA 1500 NOK
ACRO219000050 ACRO219000250
Methyl isocyanoacetate 95% tech.
Methyl isocyanoacetate
Formel: C₄H₅NO₂
MW: 99,09 g/mol
Kokepunkt: 75…76 °C (10 mmHg)
Tetthet: 1,09 g/cm³
Flash punkt: 84 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00000006
CAS-nummer: 39687-95-1
EINECS: 254-593-8
FN: 2922
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4161 to 1.4191 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR