Bis(2-methoxyethyl)phthalate ≥96.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Bis(2-methoxyethyl) phthalate

P0965-25G P0965-500G
TCIAP0965-25GEA 220 NOK
TCIAP0965-25G TCIAP0965-500G
Bis(2-methoxyethyl)phthalate ≥96.0% (by GC)
Bis(2-methoxyethyl)phthalate
Formel: C₁₄H₁₈O₆
MW: 282,29 g/mol
Kokepunkt: 340°C
Flash punkt: 185°C
MDL nummer: MFCD00042842
CAS-nummer: 117-82-8
EINECS: 204-212-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65ada4d9985fbe078ab75ddb2d29b1b6cf246ac7&vid=65ada4d9985fbe078ab75ddb2d29b1b6cf246ac7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR