2,5-Thiophenedicarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Thiophene-2,5-dicarboxylic acid

T2347-25G T2347-5G
TCIAT2347-25GEA 1325 NOK
TCIAT2347-25G TCIAT2347-5G
2,5-Thiophenedicarboxylic acid ≥98.0% (by GC, titration analysis)
2,5-Thiophenedicarboxylic acid
Formel: C₆H₄O₄S
MW: 172,16 g/mol
Smeltepunkt: 359°C
MDL nummer: MFCD00016896
CAS-nummer: 4282-31-9
EINECS: 224-284-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0db2882f89cdd4fe46804d532775aeb2ea643fd6&vid=0db2882f89cdd4fe46804d532775aeb2ea643fd6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR