Rhodium(II) acetate dimer ≥98%
Leverandør: Thermo Scientific
Synonymer: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)
L15152.04 L15152.MC L15152.ME
L15152.04EA 33800 NOK
L15152.04 L15152.MC L15152.ME
Rhodium(II) acetate dimer ≥98%
Rhodium(II) acetate dimer
Purity: 98+% CAS No.: 15956-28-2 Molecular Formula: C8H12O8Rh2 Formula Weight: 441.99 Odour: Odourless Melting Point: 205° MDL No.: MFCD00003538
Formel: C₈H₁₂O₈Rh₂
MW: 441,987 g/mol
Smeltepunkt: 205 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003538
CAS-nummer: 15956-28-2
EINECS: 240-084-8